Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCAGTACAGCTCCCCTTCTATGAA[A/G]AAGTAGCCCTTTTGCTTGAGGTTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605889 | ||||||||||||||||||||
Literature Links: |
PDLIM3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PDLIM3 - PDZ and LIM domain 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114107.4 | 979 | Silent Mutation | TTC,TTT | F,F 285 | NP_001107579.1 | |
NM_001257962.1 | 979 | Silent Mutation | TTC,TTT | F,F 245 | NP_001244891.1 | |
NM_001257963.1 | 979 | Silent Mutation | TTC,TTT | F,F 166 | NP_001244892.1 | |
NM_014476.5 | 979 | Silent Mutation | TTC,TTT | F,F 333 | NP_055291.2 |