Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATATTTCTCAAACACTATGTCAA[A/C]AAGTAGTCAGTGTAAAAAATGTTAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600951 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TERF1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TERF1 - telomeric repeat binding factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003218.3 | Intron | NP_003209.2 | ||||
NM_017489.2 | Intron | NP_059523.2 | ||||
XM_005251291.3 | Intron | XP_005251348.1 | ||||
XM_005251292.3 | Intron | XP_005251349.1 | ||||
XM_011517582.2 | Intron | XP_011515884.1 | ||||
XM_017013792.1 | Intron | XP_016869281.1 |