Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCTTCCTTTGTGTGAGACTCACT[A/T]GAGATACAGAGGTGAATGCAGTACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607738 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KCNB2 - potassium voltage-gated channel subfamily B member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004770.2 | Intron | NP_004761.2 | ||||
XM_017013981.1 | Intron | XP_016869470.1 | ||||
XM_017013982.1 | Intron | XP_016869471.1 |
Set Membership: |
HapMap |