Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAAAAGGAAGTAATCTCTGTTGC[A/G]TAGTAGTTTGAAATGGTTTCAAGAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605191 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTAF1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BTAF1 - B-TFIID TATA-box binding protein associated factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003972.2 | Intron | NP_003963.1 | ||||
XM_011540326.1 | Intron | XP_011538628.1 | ||||
XM_011540327.2 | Intron | XP_011538629.1 | ||||
XM_011540328.2 | Intron | XP_011538630.1 | ||||
XM_011540329.2 | Intron | XP_011538631.1 | ||||
XM_017016877.1 | Intron | XP_016872366.1 | ||||
XM_017016878.1 | Intron | XP_016872367.1 | ||||
XM_017016879.1 | Intron | XP_016872368.1 | ||||
XM_017016880.1 | Intron | XP_016872369.1 | ||||
XM_017016881.1 | Intron | XP_016872370.1 |