Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACCTAGCAATCATGCAAAGTCAC[A/G]GCTTTAAAGTGACCAAGAAACACTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 192974 MIM: 603708 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITGA2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ITGA2 - integrin subunit alpha 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MOCS2 - molybdenum cofactor synthesis 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004531.4 | 2955 | UTR 3 | NP_004522.1 | |||
NM_176806.3 | 2955 | UTR 3 | NP_789776.1 |
Set Membership: |
HapMap |