Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATAATCTTCAACTTGAAAGAAAA[A/C]GTCTCATGTTCAGACTGTTTAACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM126B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM126B - family with sequence similarity 126 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321618.1 | 6912 | UTR 3 | NP_001308547.1 | |||
NM_001321619.1 | 6912 | UTR 3 | NP_001308548.1 | |||
NM_001321621.1 | 6912 | UTR 3 | NP_001308550.1 | |||
NM_001321622.1 | 6912 | UTR 3 | NP_001308551.1 | |||
NM_001321623.1 | 6912 | UTR 3 | NP_001308552.1 | |||
NM_001321624.1 | 6912 | UTR 3 | NP_001308553.1 | |||
NM_001321625.1 | 6912 | UTR 3 | NP_001308554.1 | |||
NM_001321626.1 | 6912 | UTR 3 | NP_001308555.1 | |||
NM_001321627.1 | 6912 | UTR 3 | NP_001308556.1 | |||
NM_001321628.1 | 6912 | UTR 3 | NP_001308557.1 | |||
NM_001321629.1 | 6912 | UTR 3 | NP_001308558.1 | |||
NM_173822.4 | 6912 | UTR 3 | NP_776183.1 | |||
XM_006712443.3 | 6912 | Intron | XP_006712506.1 | |||
XM_011511002.2 | 6912 | Intron | XP_011509304.1 | |||
XM_017003879.1 | 6912 | Intron | XP_016859368.1 | |||
XM_017003880.1 | 6912 | Intron | XP_016859369.1 | |||
XM_017003881.1 | 6912 | Intron | XP_016859370.1 | |||
XM_017003882.1 | 6912 | Intron | XP_016859371.1 | |||
XM_017003883.1 | 6912 | Intron | XP_016859372.1 |