Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCAGAAGTCAGGCTTTCCACAGA[C/G]AGACGGGACTACATGGCCACCTGGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300520 MIM: 300575 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLDN2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CLDN2 - claudin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171092.1 | 131 | UTR 5 | NP_001164563.1 | |||
NM_001171095.1 | 131 | Intron | NP_001164566.1 | |||
NM_020384.3 | 131 | Intron | NP_065117.1 |
RIPPLY1 - ripply transcriptional repressor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171706.1 | 131 | UTR 3 | NP_001165177.1 | |||
NM_138382.2 | 131 | UTR 3 | NP_612391.1 |
Set Membership: |
HapMap |