Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAATATTAGTGCTGAAGAAGCACA[A/G]TTGTTTGGAAAAATACAGGACCCAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613166 MIM: 606453 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCLK2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DCLK2 - doublecortin like kinase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRBA - LPS responsive beige-like anchor protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199282.2 | 9482 | UTR 3 | NP_001186211.2 | |||
NM_006726.4 | 9482 | UTR 3 | NP_006717.2 | |||
XM_005263372.3 | 9482 | UTR 3 | XP_005263429.1 | |||
XM_005263373.2 | 9482 | UTR 3 | XP_005263430.1 | |||
XM_005263374.3 | 9482 | UTR 3 | XP_005263431.1 | |||
XM_005263375.3 | 9482 | UTR 3 | XP_005263432.1 | |||
XM_011532434.2 | 9482 | UTR 3 | XP_011530736.1 | |||
XM_017008872.1 | 9482 | UTR 3 | XP_016864361.1 | |||
XM_017008873.1 | 9482 | UTR 3 | XP_016864362.1 | |||
XM_017008874.1 | 9482 | UTR 3 | XP_016864363.1 |