Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGGCTCTGGAATTCTCTCCAACA[A/C]CAAATCTGTGGCTTTCCCATTATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
41 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610115 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NDC1 - NDC1 transmembrane nucleoporin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168551.1 | Intron | NP_001162023.1 | ||||
NM_018087.4 | Intron | NP_060557.3 | ||||
XM_011541766.2 | Intron | XP_011540068.1 |