Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGACACAACACCATCCAAAACCC[A/G]TGGTCTCTTCATTCAGATACCCACA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 306480 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PUDP PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PUDP - pseudouridine 5'-phosphatase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135565.1 | Intron | NP_001129037.1 | ||||
NM_001178135.1 | Intron | NP_001171606.1 | ||||
NM_001178136.1 | Intron | NP_001171607.1 | ||||
NM_012080.4 | Intron | NP_036212.3 |
Set Membership: |
HapMap |