Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGCTCTTTCTCTTACTTTCCTCG[A/G]CTTAGAATATAGTTTTATTACATTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
BBOF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BBOF1 - basal body orientation factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025057.2 | Intron | NP_079333.2 | ||||
XM_005268092.3 | Intron | XP_005268149.1 | ||||
XM_005268097.3 | Intron | XP_005268154.1 | ||||
XM_005268098.4 | Intron | XP_005268155.1 | ||||
XM_011537170.2 | Intron | XP_011535472.1 | ||||
XM_011537171.2 | Intron | XP_011535473.1 | ||||
XM_011537172.1 | Intron | XP_011535474.1 | ||||
XM_011537174.2 | Intron | XP_011535476.1 | ||||
XM_011537175.2 | Intron | XP_011535477.1 | ||||
XM_011537176.2 | Intron | XP_011535478.1 | ||||
XM_011537177.2 | Intron | XP_011535479.1 | ||||
XM_011537178.2 | Intron | XP_011535480.1 | ||||
XM_011537179.2 | Intron | XP_011535481.1 | ||||
XM_017021659.1 | Intron | XP_016877148.1 | ||||
XM_017021660.1 | Intron | XP_016877149.1 | ||||
XM_017021661.1 | Intron | XP_016877150.1 | ||||
XM_017021662.1 | Intron | XP_016877151.1 | ||||
XM_017021663.1 | Intron | XP_016877152.1 |