Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCGTCGGCCGGGCACTGGTGTCCC[A/G]TGTCACTCAGCGGAGGGAGGCTGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606102 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CACTIN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CACTIN - cactin, spliceosome C complex subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIP5K1C - phosphatidylinositol-4-phosphate 5-kinase type 1 gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195733.1 | 4101 | UTR 3 | NP_001182662.1 | |||
NM_001300849.1 | 4101 | Intron | NP_001287778.1 | |||
NM_012398.2 | 4101 | UTR 3 | NP_036530.1 | |||
XM_011527845.1 | 4101 | Intron | XP_011526147.1 | |||
XM_011527846.1 | 4101 | Intron | XP_011526148.1 | |||
XM_011527848.1 | 4101 | Intron | XP_011526150.1 | |||
XM_011527849.1 | 4101 | Intron | XP_011526151.1 | |||
XM_011527850.2 | 4101 | Intron | XP_011526152.1 | |||
XM_011527851.2 | 4101 | Intron | XP_011526153.1 | |||
XM_011527853.2 | 4101 | Intron | XP_011526155.1 | |||
XM_017026540.1 | 4101 | UTR 3 | XP_016882029.1 |