Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGTCACCAGCTTTGTTTCCTTAA[A/G]TCCAAAACTTTGGAAATAAATTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608951 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNOT6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CNOT6 - CCR4-NOT transcription complex subunit 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303241.1 | Intron | NP_001290170.1 | ||||
XM_005265953.1 | Intron | XP_005266010.1 | ||||
XM_011534605.2 | Intron | XP_011532907.1 | ||||
XM_011534606.1 | Intron | XP_011532908.1 | ||||
XM_011534607.1 | Intron | XP_011532909.1 | ||||
XM_017009670.1 | Intron | XP_016865159.1 | ||||
XM_017009671.1 | Intron | XP_016865160.1 | ||||
XM_017009672.1 | Intron | XP_016865161.1 |
Set Membership: |
HapMap |