Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACACCAAATAATCATTTGATATTA[A/G]GTGCTTTACATTGTAAATGAGCTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DTWD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DTWD2 - DTW domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308081.1 | 3976 | UTR 3 | NP_001295010.1 | |||
NM_173666.3 | 3976 | UTR 3 | NP_775937.1 | |||
XM_011543338.2 | 3976 | UTR 3 | XP_011541640.2 | |||
XM_011543340.2 | 3976 | UTR 3 | XP_011541642.1 |