Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATACTGCTCTCATAATTCTTTACT[A/C]AACATTTGGAGGGTTTTAATAGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC82 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC82 - coiled-coil domain containing 82 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318736.1 | 4437 | Intron | NP_001305665.1 | |||
NM_001318737.1 | 4437 | Intron | NP_001305666.1 | |||
NM_024725.3 | 4437 | Intron | NP_079001.2 | |||
XM_005274292.3 | 4437 | Intron | XP_005274349.1 | |||
XM_006718911.3 | 4437 | Intron | XP_006718974.1 | |||
XM_011542984.2 | 4437 | Intron | XP_011541286.1 | |||
XM_011542987.2 | 4437 | Intron | XP_011541289.1 | |||
XM_011542988.2 | 4437 | Intron | XP_011541290.1 | |||
XM_017018305.1 | 4437 | Intron | XP_016873794.1 | |||
XM_017018306.1 | 4437 | Intron | XP_016873795.1 | |||
XM_017018307.1 | 4437 | Intron | XP_016873796.1 | |||
XM_017018308.1 | 4437 | Intron | XP_016873797.1 | |||
XM_017018309.1 | 4437 | UTR 3 | XP_016873798.1 | |||
XM_017018310.1 | 4437 | UTR 3 | XP_016873799.1 | |||
XM_017018311.1 | 4437 | Intron | XP_016873800.1 | |||
XM_017018312.1 | 4437 | Intron | XP_016873801.1 |