Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGACAAGACCGAGCCTAAGGCCTA[C/T]AGGCGGCGGCGGTCCCTCAGCCCAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603309 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK13 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CDK13 - cyclin dependent kinase 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003718.4 | 2071 | Silent Mutation | TAC,TAT | Y,Y 310 | NP_003709.3 | |
NM_031267.3 | 2071 | Silent Mutation | TAC,TAT | Y,Y 310 | NP_112557.2 | |
XM_011515597.2 | 2071 | Silent Mutation | TAC,TAT | Y,Y 310 | XP_011513899.1 | |
XM_017012750.1 | 2071 | Silent Mutation | TAC,TAT | Y,Y 310 | XP_016868239.1 | |
XM_017012751.1 | 2071 | Silent Mutation | TAC,TAT | Y,Y 310 | XP_016868240.1 |
Set Membership: |
HapMap |