Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTGGAAACAGGTTGAGGCTTATT[C/T]CGAAGACCTCCCTAATCTTCTGCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300163 MIM: 300930 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FHL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FHL1 - four and a half LIM domains 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAP7D3 - MAP7 domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173516.1 | Intron | NP_001166987.1 | ||||
NM_001173517.1 | Intron | NP_001166988.1 | ||||
NM_024597.3 | Intron | NP_078873.2 | ||||
XM_005262472.1 | Intron | XP_005262529.1 | ||||
XM_017029843.1 | Intron | XP_016885332.1 |
Set Membership: |
HapMap |