Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGAAGTTCTGAGCTTTGCCTAGA[A/G]AACACCCTTTGGCGGGAGAGCTGGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
39 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MORN1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MORN1 - MORN repeat containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RER1 - retention in endoplasmic reticulum sorting receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007033.4 | Intron | NP_008964.3 | ||||
XM_005244713.4 | Intron | XP_005244770.2 | ||||
XM_006710306.2 | Intron | XP_006710369.1 | ||||
XM_011540542.1 | Intron | XP_011538844.1 | ||||
XM_011540543.1 | Intron | XP_011538845.1 | ||||
XM_017000131.1 | Intron | XP_016855620.1 | ||||
XM_017000132.1 | Intron | XP_016855621.1 | ||||
XM_017000133.1 | Intron | XP_016855622.1 | ||||
XM_017000134.1 | Intron | XP_016855623.1 | ||||
XM_017000135.1 | Intron | XP_016855624.1 |