Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATGAAACAAGTATGTCTTTATA[G/T]TAAAGTATTTTAGGGTCTGGTAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600562 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDH12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CDH12 - cadherin 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317227.1 | Intron | NP_001304156.1 | ||||
NM_001317228.1 | Intron | NP_001304157.1 | ||||
NM_004061.4 | Intron | NP_004052.2 | ||||
XM_011513927.2 | Intron | XP_011512229.1 | ||||
XM_017008920.1 | Intron | XP_016864409.1 | ||||
XM_017008921.1 | Intron | XP_016864410.1 |
Set Membership: |
HapMap |