Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTAGACGCTCTCCAGGCTATTGCT[A/G]CCTCATACAGAGATACCATGTTTGC
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||
Phenotype: |
MIM: 608113 | ||||||||||||||||||||||||||
Literature Links: |
SGCZ PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
SGCZ - sarcoglycan zeta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322879.1 | 5694 | UTR 3 | NP_001309808.1 | |||
NM_001322880.1 | 5694 | UTR 3 | NP_001309809.1 | |||
NM_001322881.1 | 5694 | UTR 3 | NP_001309810.1 | |||
NM_139167.3 | 5694 | UTR 3 | NP_631906.2 |
Set Membership: |
HapMap |