Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAACATGAAATGACCTAGAAGCA[C/G]TGTTACAAATCTTACCCCAGAAATA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612753 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCBE1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CCBE1 - collagen and calcium binding EGF domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_133459.3 | 4766 | UTR 3 | NP_597716.1 | |||
XM_017025556.1 | 4766 | Intron | XP_016881045.1 | |||
XM_017025557.1 | 4766 | Intron | XP_016881046.1 | |||
XM_017025558.1 | 4766 | Intron | XP_016881047.1 |
Set Membership: |
HapMap |