Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATAGTAGGGTTCATGTCTAAAAT[C/T]TTCCCTTGGGGTGGGAGTAGGTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611125 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DSEL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DSEL - dermatan sulfate epimerase-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032160.2 | 7388 | UTR 3 | NP_115536.1 |
LOC643542 - uncharacterized LOC643542 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |