Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164730 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AKT1 - AKT serine/threonine kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014431.1 | Intron | NP_001014431.1 | ||||
NM_001014432.1 | Intron | NP_001014432.1 | ||||
NM_005163.2 | Intron | NP_005154.2 | ||||
XM_005267401.1 | Intron | XP_005267458.1 | ||||
XM_017021075.1 | Intron | XP_016876564.1 | ||||
XM_017021076.1 | Intron | XP_016876565.1 | ||||
XM_017021077.1 | Intron | XP_016876566.1 | ||||
XM_017021078.1 | Intron | XP_016876567.1 |
Set Membership: |
HapMap |