Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTCCTTGTTTTAGGCATATCCTG[A/G]GCAACTATCCTTGTGATACCGCCCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614277 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBE2W PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
UBE2W - ubiquitin conjugating enzyme E2 W (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001481.2 | Intron | NP_001001481.2 | ||||
NM_001271015.1 | Intron | NP_001257944.1 | ||||
NM_018299.4 | Intron | NP_060769.4 |
Set Membership: |
HapMap |