Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAGGGAAACCTGTTGTCTGTGTT[C/T]AGAGTGGGTACCTGGCTTACTGACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
25 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605551 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOS1AP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NOS1AP - nitric oxide synthase 1 adaptor protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001126060.1 | Intron | NP_001119532.2 | ||||
NM_001164757.1 | Intron | NP_001158229.1 | ||||
NM_014697.2 | Intron | NP_055512.1 |
Set Membership: |
HapMap |