Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCAAACTGATCAGTCTTTGGTTTA[C/G]TTTGGTTTGACTTTTTTCATATACA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C4orf19 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C4orf19 - chromosome 4 open reading frame 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001104629.1 | Intron | NP_001098099.1 | ||||
NM_018302.2 | Intron | NP_060772.2 | ||||
XM_011513712.2 | Intron | XP_011512014.1 | ||||
XM_011513713.2 | Intron | XP_011512015.1 | ||||
XM_017008361.1 | Intron | XP_016863850.1 |