Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTGACCACAGGAGTCTCCCAGAA[A/G]CAGGGCTTTATTTTATGTCTCACAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182098 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SCTR PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SCTR - secretin receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM37 - transmembrane protein 37 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_183240.2 | Intron | NP_899063.2 | ||||
XM_005263598.4 | Intron | XP_005263655.1 | ||||
XM_006712300.3 | Intron | XP_006712363.1 | ||||
XM_011510659.2 | Intron | XP_011508961.1 |
Set Membership: |
HapMap |