Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGTAAGAAATAAGCATCCTAGCCG[A/C]GACTTGATTATTTGGTTCAAATATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600153 MIM: 605857 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100506142 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC100506142 - uncharacterized LOC100506142 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIGF - phosphatidylinositol glycan anchor biosynthesis class F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002643.3 | Intron | NP_002634.1 | ||||
NM_173074.2 | Intron | NP_775097.1 | ||||
XM_005264369.2 | Intron | XP_005264426.1 | ||||
XM_011532908.2 | Intron | XP_011531210.1 |
RHOQ - ras homolog family member Q | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |