Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATTATTTTAAATGTAACAACTAC[C/T]CCTCAATATAAGATGATGAGTAGAA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 606400 | |||||||||||||||||||||||
Literature Links: |
CAPN7 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CAPN7 - calpain 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014296.2 | Intron | NP_055111.1 | ||||
XM_011533555.2 | Intron | XP_011531857.1 | ||||
XM_011533556.2 | Intron | XP_011531858.1 | ||||
XM_011533557.2 | Intron | XP_011531859.1 | ||||
XM_017006043.1 | Intron | XP_016861532.1 | ||||
XM_017006044.1 | Intron | XP_016861533.1 | ||||
XM_017006045.1 | Intron | XP_016861534.1 |