Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAATGTCCTTAAAAGCTGTTAGTT[A/C]TATAGGGTTTAATGAGCCACAAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611241 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPRIN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPRIN3 - GPRIN family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198281.2 | 5398 | UTR 3 | NP_938022.2 | |||
XM_005262936.3 | 5398 | UTR 3 | XP_005262993.1 | |||
XM_005262937.4 | 5398 | UTR 3 | XP_005262994.1 | |||
XM_017008043.1 | 5398 | UTR 3 | XP_016863532.1 | |||
XM_017008044.1 | 5398 | UTR 3 | XP_016863533.1 |