Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTAAGACAAATAATACTTTCCTCA[A/G]ATAGTTCAGAAAATTTTTAAAAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102581 MIM: 603056 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACVR2A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ACVR2A - activin A receptor type 2A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ORC4 - origin recognition complex subunit 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190879.2 | 3413 | UTR 3 | NP_001177808.1 | |||
NM_001190881.2 | 3413 | Intron | NP_001177810.1 | |||
NM_001190882.2 | 3413 | UTR 3 | NP_001177811.1 | |||
NM_002552.4 | 3413 | UTR 3 | NP_002543.2 | |||
NM_181741.3 | 3413 | UTR 3 | NP_859525.1 | |||
NM_181742.3 | 3413 | Intron | NP_859526.1 | |||
XM_011511255.2 | 3413 | Intron | XP_011509557.1 | |||
XM_017004226.1 | 3413 | Intron | XP_016859715.1 | |||
XM_017004227.1 | 3413 | Intron | XP_016859716.1 |
Set Membership: |
HapMap |