Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAGTCACTGGGTTTTGTTCTTGCA[A/C]CTTGAGTTTCTTTTGGCTTTGCCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604865 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLF7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KLF7 - Kruppel like factor 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270942.1 | 5171 | UTR 3 | NP_001257871.1 | |||
NM_001270943.1 | 5171 | UTR 3 | NP_001257872.1 | |||
NM_001270944.1 | 5171 | UTR 3 | NP_001257873.1 | |||
NM_003709.3 | 5171 | UTR 3 | NP_003700.1 | |||
XM_005246926.3 | 5171 | UTR 3 | XP_005246983.1 | |||
XM_006712815.3 | 5171 | UTR 3 | XP_006712878.1 | |||
XM_011512072.2 | 5171 | UTR 3 | XP_011510374.1 | |||
XM_011512073.2 | 5171 | UTR 3 | XP_011510375.1 | |||
XM_011512074.2 | 5171 | UTR 3 | XP_011510376.1 | |||
XM_011512075.1 | 5171 | Intron | XP_011510377.1 | |||
XM_017005161.1 | 5171 | UTR 3 | XP_016860650.1 | |||
XM_017005162.1 | 5171 | UTR 3 | XP_016860651.1 | |||
XM_017005163.1 | 5171 | Intron | XP_016860652.1 | |||
XM_017005164.1 | 5171 | UTR 3 | XP_016860653.1 |
Set Membership: |
HapMap |