Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGTGCTCTAACTCCAGAGCATT[C/T]TGCATACATCTTTAGGGTTATTCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610032 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TNPO3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TNPO3 - transportin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001191028.2 | Intron | NP_001177957.2 | ||||
NM_012470.3 | Intron | NP_036602.1 | ||||
XM_011515989.2 | Intron | XP_011514291.1 |
TPI1P2 - triosephosphate isomerase 1 pseudogene 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |