Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGTGATAAGCCACCAGGGTTCTGT[C/T]GGGAAGCCTAGGACAGGCCATTTTA
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||
Phenotype: |
MIM: 302910 | ||||||||||||||||||||||||||
Literature Links: |
CLCN4 PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CLCN4 - chloride voltage-gated channel 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256944.1 | Intron | NP_001243873.1 | ||||
NM_001830.3 | Intron | NP_001821.2 |
Set Membership: |
HapMap |