Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTGGCACGGCTGCTGGCTCGCCA[C/T]CCGCGCTGTGTGCCCGACGGACCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCTD21 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCTD21 - potassium channel tetramerization domain containing 21 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP35 - ubiquitin specific peptidase 35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020798.2 | 806 | Silent Mutation | CAC,CAT | H,H 151 | NP_065849.1 | |
XM_011545187.2 | 806 | Intron | XP_011543489.1 | |||
XM_011545188.2 | 806 | Intron | XP_011543490.1 | |||
XM_017018052.1 | 806 | Silent Mutation | CAC,CAT | H,H 151 | XP_016873541.1 | |
XM_017018053.1 | 806 | Intron | XP_016873542.1 |