Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACTGAAGTCCTAGGTAGCCAACC[A/G]TAGCTACTGGGGTGCAAGAAGAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602855 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSMA6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PSMA6 - proteasome subunit alpha 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282232.1 | Intron | NP_001269161.1 | ||||
NM_001282233.1 | Intron | NP_001269162.1 | ||||
NM_001282234.1 | Intron | NP_001269163.1 | ||||
NM_002791.2 | Intron | NP_002782.1 | ||||
XM_017021468.1 | Intron | XP_016876957.1 |
Set Membership: |
HapMap |