Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGGGATCATGTAGCCCGGGTTGCC[C/G]GGGCTGGGGTGCGAGTGCGTGTGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604974 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101927248 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC101927248 - uncharacterized LOC101927248 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SOX21 - SRY-box 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007084.3 | 1181 | Silent Mutation | CCC,CCG | P,P 229 | NP_009015.1 |
SOX21-AS1 - SOX21 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |