Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAACAAATAAGATGCTTTACATCT[A/G]TCAGCTCAGAGTCTGCTGTTATTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610404 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RMI1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RMI1 - RecQ mediated genome instability 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024945.2 | Intron | NP_079221.2 | ||||
XM_005252211.2 | Intron | XP_005252268.1 | ||||
XM_005252213.2 | Intron | XP_005252270.1 | ||||
XM_011519034.2 | Intron | XP_011517336.1 | ||||
XM_017015140.1 | Intron | XP_016870629.1 |
Set Membership: |
HapMap |