Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTCACTTTTTTTGTATGTGTGGA[A/G]GAGGAGAACTGGTGTGGGAAAATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614517 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARNTL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARNTL2 - aryl hydrocarbon receptor nuclear translocator like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001248002.1 | Intron | NP_001234931.1 | ||||
NM_001248003.1 | Intron | NP_001234932.1 | ||||
NM_001248004.1 | Intron | NP_001234933.1 | ||||
NM_001248005.1 | Intron | NP_001234934.1 | ||||
NM_020183.4 | Intron | NP_064568.3 | ||||
XM_006719112.3 | Intron | XP_006719175.1 | ||||
XM_006719114.3 | Intron | XP_006719177.1 | ||||
XM_011520766.2 | Intron | XP_011519068.1 | ||||
XM_011520768.2 | Intron | XP_011519070.1 | ||||
XM_011520769.2 | Intron | XP_011519071.1 | ||||
XM_011520770.2 | Intron | XP_011519072.1 | ||||
XM_011520771.2 | Intron | XP_011519073.1 | ||||
XM_017019666.1 | Intron | XP_016875155.1 | ||||
XM_017019667.1 | Intron | XP_016875156.1 | ||||
XM_017019668.1 | Intron | XP_016875157.1 | ||||
XM_017019669.1 | Intron | XP_016875158.1 |
Set Membership: |
HapMap |