Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACCTGGCTATGCCTGTTTAATCC[C/T]ACTCTCCTGGCTGGGAGACAAGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300684 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
XKRX PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
XKRX - XK related, X-linked | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_212559.2 | Intron | NP_997724.2 | ||||
XM_005262130.2 | Intron | XP_005262187.1 | ||||
XM_011530954.2 | Intron | XP_011529256.1 | ||||
XM_011530955.1 | Intron | XP_011529257.1 | ||||
XM_017029517.1 | Intron | XP_016885006.1 |