Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCACAGTAGCCAAAGATACACGTT[C/T]ACAAAAGTCTTCATCTTTGAGAGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601484 | ||||||||||||||||||||
Literature Links: |
CCDC152 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC152 - coiled-coil domain containing 152 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134848.1 | 2361 | UTR 3 | NP_001128320.1 |
SEPP1 - selenoprotein P, plasma, 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085486.1 | 2361 | Missense Mutation | AAA,GAA | K,E 187 | NP_001078955.1 | |
NM_001093726.1 | 2361 | Missense Mutation | AAA,GAA | K,E 217 | NP_001087195.1 | |
NM_005410.2 | 2361 | Missense Mutation | AAA,GAA | K,E 187 | NP_005401.3 |