Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATCTGGGAAGAAGCTGTGTCAGA[A/C]GAACCAGGCAGGCCATTATATGACA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
34 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602458 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYBPHL PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MYBPHL - myosin binding protein H like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SORT1 - sortilin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001205228.1 | Intron | NP_001192157.1 | ||||
NM_002959.6 | Intron | NP_002950.3 | ||||
XM_005271100.2 | Intron | XP_005271157.1 | ||||
XM_005271101.2 | Intron | XP_005271158.1 | ||||
XM_005271102.1 | Intron | XP_005271159.1 | ||||
XM_006710812.1 | Intron | XP_006710875.1 |
Set Membership: |
HapMap |