Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATCAGTCTCGGGCTTCTTTGCTG[C/G]AGGGGTCGTGAGCCCTCAAACAGGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606094 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEACAM18 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CEACAM18 - carcinoembryonic antigen related cell adhesion molecule 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278392.1 | Intron | NP_001265321.1 | ||||
XM_011527258.2 | Intron | XP_011525560.1 |
SIGLEC12 - sialic acid binding Ig like lectin 12 (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033329.2 | Intron | NP_201586.1 | ||||
NM_053003.3 | Intron | NP_443729.1 | ||||
XM_011527441.2 | Intron | XP_011525743.1 | ||||
XM_011527442.2 | Intron | XP_011525744.1 |
Set Membership: |
HapMap |