Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCCACTTGTAACCACAAAAAAGC[A/G]AGCTCACAGCATTAAGTGCCTAATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614394 MIM: 612912 MIM: 191161 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFT20 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IFT20 - intraflagellar transport 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267774.1 | Intron | NP_001254703.1 | ||||
NM_001267775.1 | Intron | NP_001254704.1 | ||||
NM_001267776.1 | Intron | NP_001254705.1 | ||||
NM_001267777.1 | Intron | NP_001254706.1 | ||||
NM_001267778.1 | Intron | NP_001254707.1 | ||||
NM_174887.3 | Intron | NP_777547.1 |
TMEM97 - transmembrane protein 97 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNFAIP1 - TNF alpha induced protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |