Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTTGTCTCTTGTCCATATAGTGC[A/G]TTCCTCGGAGTAGATGGCCAGTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609030 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DGCR8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DGCR8 - DGCR8 microprocessor complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190326.1 | Intron | NP_001177255.1 | ||||
NM_022720.6 | Intron | NP_073557.3 | ||||
XM_006724268.2 | Intron | XP_006724331.1 |
MIR1306 - microRNA 1306 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3618 - microRNA 3618 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |