Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607629 MIM: 604832 MIM: 615782 | ||||||||||||||||||||
Literature Links: |
APH1A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
APH1A - aph-1 homolog A, gamma-secretase subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C1orf54 - chromosome 1 open reading frame 54 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CA14 - carbonic anhydrase 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CIART - circadian associated repressor of transcription | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |