Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAAGAGCAGGGCCTGTTGTCCGGC[C/T]CCACAATTTCCTTCCAGGTGGAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179780 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPEP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DPEP1 - dipeptidase 1 (renal) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128141.2 | Intron | NP_001121613.1 | ||||
NM_004413.3 | Intron | NP_004404.1 | ||||
XM_005256285.4 | Intron | XP_005256342.1 | ||||
XM_005256286.3 | Intron | XP_005256343.1 | ||||
XM_011522925.2 | Intron | XP_011521227.1 | ||||
XM_017023009.1 | Intron | XP_016878498.1 |
Set Membership: |
HapMap |