Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAATACTCTTGAGACAGTGTAGGA[A/C]TTTGAAGTGGTTGACTGGATCAAGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609664 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NLRP11 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NLRP11 - NLR family pyrin domain containing 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297743.1 | 3121 | UTR 3 | NP_001284672.1 | |||
NM_145007.3 | 3121 | UTR 3 | NP_659444.2 |