Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTTCCAGGGATATTTGTGGGAAA[C/G]CAGTACACAATAATATATATGGAAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 126065 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP24A1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CYP24A1 - cytochrome P450 family 24 subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000782.4 | Intron | NP_000773.2 | ||||
NM_001128915.1 | Intron | NP_001122387.1 | ||||
XM_005260304.4 | Intron | XP_005260361.1 | ||||
XM_017027691.1 | Intron | XP_016883180.1 | ||||
XM_017027692.1 | Intron | XP_016883181.1 | ||||
XM_017027693.1 | Intron | XP_016883182.1 |
Set Membership: |
HapMap |