Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAAAGGAAAAACAGATAAAAGTCA[G/T]CTTTGGGAATGGGCCTCATAGGTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 126141 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPP6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DPP6 - dipeptidyl peptidase like 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039350.2 | Intron | NP_001034439.1 | ||||
NM_001290252.1 | Intron | NP_001277181.1 | ||||
NM_001290253.1 | Intron | NP_001277182.1 | ||||
NM_001936.4 | Intron | NP_001927.3 | ||||
NM_130797.3 | Intron | NP_570629.2 | ||||
XM_017011808.1 | Intron | XP_016867297.1 | ||||
XM_017011809.1 | Intron | XP_016867298.1 | ||||
XM_017011810.1 | Intron | XP_016867299.1 | ||||
XM_017011811.1 | Intron | XP_016867300.1 | ||||
XM_017011812.1 | Intron | XP_016867301.1 |
Set Membership: |
HapMap |